Iridium ≥99.99% (trace metals basis), powder -22 mesh

Supplier: BeanTown Chemical

132120-1G 132120-5G
BT132120-1GEA 510.48 USD
BT132120-1G BT132120-5G
Iridium ≥99.99% (trace metals basis), powder -22 mesh
Iridium

CAS: 7439-88-5; EC No: 231-095-9; MDL No: MFCD00011062 Powder; Molecular Formula: Ir; MW: 192.22 Melting Point: 2450°; Boiling Point: 4130° Density (g/mL): 22.65


  • Powder

Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 4130 °C
Melting Pt: 2450 °C
Density: 22.65 g/mL
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR