6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one

132115-1G 132115-250MG 132115-5G
BT132115-1GEA 69 USD
BT132115-1G BT132115-250MG BT132115-5G
6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥98%
6-Methyl-4-trifluoromethyl-2(1H)-pyridone

CAS: 22123-19-9; MDL No: MFCD00042479 Powder; Molecular Formula: C7H6F3NO; MW: 177.13 Melting Point: 129-130°


  • Powder

Formula: C₇H₆F₃NO
MW: 177.126 g/mol
Melting Pt: 129…130 °C
MDL Number: MFCD00042479
CAS Number: 22123-19-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR