Zirconium disulphate tetrahydrate ≥98%
Supplier: BeanTown Chemical
132080-100G
132080-2KG
132080-500G
BT132080-100GEA
36.25
USD
BT132080-100G
BT132080-2KG
BT132080-500G
Zirconium disulphate tetrahydrate ≥98%
Zirconium disulphate tetrahydrate
CAS: 7446-31-3; EC No: 238-694-4; MDL No: MFCD00149267
-20 Mesh Powder; Linear Formula: Zr(SO4)2·4H2O ; MW: 355.40
Melting Point: 410°
Density (g/mL): 3.22
- -20 Mesh Powder
Formula:
O₈S₂Zr·4H₂O MW: 355.396 g/mol Melting Pt: 410 °C Density: 3.22 g/mL |
MDL Number:
MFCD00149267 CAS Number: 7446-31-3 EINECS: 238-694-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...