Tris acetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate

132065-100G 132065-250G 132065-25G
BT132065-100GEA 128 USD
BT132065-100G BT132065-250G BT132065-25G
Tris acetate ≥99%
Tris acetate

CAS: 6850-28-8; EC No: 229-939-6; MDL No: MFCD00038951 Powder; Linear Formula: NH2C(CH2OH)3·CH3COOH; Molecular Formula: C4H11NO3·C2H4O2; MW: 181.19


  • Powder

Formula: NH₂C(CH₂OH)₃·CH₃COOH
MW: 181.1870 g/mol
MDL Number: MFCD00038951
CAS Number: 6850-28-8
EINECS: 229-939-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR