Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Supplier: BeanTown Chemical

132050-1G 132050-5G
BT132050-1GEA 616 USD
BT132050-1G BT132050-5G
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate

CAS: 35138-22-8; MDL No: MFCD00075045 Powder; Molecular Formula: C16H24BF4Rh; MW: 406.07 Melting Point: 190° (decomposes)


  • Powder

Formula: C₁₆H₂₄BF₄Rh
Melting Pt: 190 °C (dec.)
MDL Number: MFCD00075045
CAS Number: 35138-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR