Disodium hexachloroiridate hexahydrate

Supplier: BeanTown Chemical

Synonyms: Sodium hexachloroiridate(IV) hexahydrate

132020-1G 132020-5G
BT132020-1GEA 97.06 USD
BT132020-1G BT132020-5G
Disodium hexachloroiridate hexahydrate
Disodium hexachloroiridate hexahydrate

CAS: 19567-78-3; EC No: 241-011-2; MDL No: MFCD00149169 Powder; Linear Formula: Na2IrCl6·6H2O; MW: 558.99 Melting Point: 600° (decomposes)


  • Powder

Formula: Cl₆IrNa₂·6H₂O
Melting Pt: 600 °C (dec.)
MDL Number: MFCD00149169
CAS Number: 19567-78-3
EINECS: 241-011-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR