2,4,6-Trimethyl-4-heptanol ≥97%

Supplier: BeanTown Chemical

131990-1G 131990-25G 131990-5G
BT131990-1GEA 26 USD
BT131990-1G BT131990-25G BT131990-5G
2,4,6-Trimethyl-4-heptanol ≥97%
2,4,6-Trimethyl-4-heptanol

CAS: 60836-07-9; MDL No: MFCD00048354 Liquid; Molecular Formula: C10H22O; MW: 158.28


  • Liquid

Formula: C₁₀H₂₂O
MDL Number: MFCD00048354
CAS Number: 60836-07-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR