4-Propoxybenzaldehyde ≥97%

Supplier: BeanTown Chemical

131980-25G 131980-5G
BT131980-25GEA 77 USD
BT131980-25G BT131980-5G
4-Propoxybenzaldehyde ≥97%
4-Propoxybenzaldehyde

CAS: 5736-85-6; MDL No: MFCD00014134; RTECS: CU7700000 Liquid; Molecular Formula: C10H12O2 ; MW: 164.20 Boiling Point: 129-130°/10 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.039; Refractive Index: 1.546 Air Sensitive


  • RTECS: CU7700000
  • Refractive index: 1.546
  • Air Sensitive
  • Liquid

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 129…130 °C/10 mmHg
Density: 1.039 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00014134
CAS Number: 5736-85-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR