1,6-Bis(diphenylphosphino)hexane ≥97%

Supplier: BeanTown Chemical

131965-1G 131965-25G 131965-5G
BT131965-1GEA 27 USD
BT131965-1G BT131965-25G BT131965-5G
1,6-Bis(diphenylphosphino)hexane ≥97%
1,6-Bis(diphenylphosphino)hexane

CAS: 19845-69-3; MDL No: MFCD00003053 Powder; Linear Formula: (C6H5)2P(CH2)6P(C5H5)2; Molecular Formula: C30H32P2; MW: 454.52 Melting Point: 124-126°


  • Powder

Formula: C₃₀H₃₂P₂
Melting Pt: 124…126 °C
MDL Number: MFCD00003053
CAS Number: 19845-69-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR