Titanyl(IV) acetylacetonate
Supplier: BeanTown Chemical
131960-10G
131960-250G
131960-50G
BT131960-10GEA
65
USD
BT131960-10G
BT131960-250G
BT131960-50G
Titanyl(IV) acetylacetonate
Titanyl(IV) acetylacetonate
CAS: 14024-64-7; EC No: 237-861-9; MDL No: MFCD00013505; RTECS: XR2330000
Powder; Linear Formula: OTi(C5H7O2)2; MW: 262.10
Melting Point: 184° (decomposes)
- RTECS: XR2330000
- Powder
Formula:
C₁₀H₁₄O₅Ti Melting Pt: 184 °C (dec.) |
MDL Number:
MFCD00013505 CAS Number: 14024-64-7 EINECS: 237-861-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...