2,3,4-Trifluorobenzoic acid ≥98%
Supplier: BeanTown Chemical
131950-1G
131950-25G
131950-5G
BT131950-1GEA
34
USD
BT131950-1G
BT131950-25G
BT131950-5G
2,3,4-Trifluorobenzoic acid ≥98%
2,3,4-Trifluorobenzoic acid
CAS: 61079-72-9; MDL No: MFCD00061232
Powder; Molecular Formula: C7H3F3O2; MW: 176.09
Melting Point: 140-142°
- Powder
Formula:
C₇H₃F₃O₂ Melting Pt: 140…142 °C |
MDL Number:
MFCD00061232 CAS Number: 61079-72-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...