5,5-Dimethylhydantoin ≥97%
Supplier: BeanTown Chemical
Synonyms:
5,5-Dimethylimidazolidine-2,4-dione
131930-100G
131930-500G
BT131930-100GEA
38
USD
BT131930-100G
BT131930-500G
5,5-Dimethylhydantoin ≥97%
5,5-Dimethylhydantoin
CAS: 77-71-4; EC No: 201-051-3; MDL No: MFCD00005266; RTECS: MU0977000
Crystalline/Powder; Molecular Formula: C5H8N2O2; MW: 128.13
Melting Point: 174-177°
- RTECS: MU0977000
- Crystalline/Powder
Formula:
C₅H₈N₂O₂ Melting Pt: 174…177 °C |
MDL Number:
MFCD00005266 CAS Number: 77-71-4 EINECS: 201-051-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...