1-(1-Adamantyl)ethylamine hydrochloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Rimantadine Hydrochloride, alpha-Methyl-1-adamantanemethylamine Hydrochloride

131910-25G 131910-5G
BT131910-25GEA 269 USD
BT131910-25G BT131910-5G
1-(1-Adamantyl)ethylamine hydrochloride ≥99%
1-(1-Adamantyl)ethylamine hydrochloride

CAS: 1501-84-4; MDL No: MFCD00072023; RTECS: AU4717000 Crystalline; Molecular Formula: C12H22ClN; MW: 215.76


  • RTECS: AU4717000
  • Crystalline

Formula: C₁₂H₂₁N
MDL Number: MFCD00072023
CAS Number: 1501-84-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR