Lithium metaborate 98.0-102.0% ACS

Supplier: BeanTown Chemical

131865-250G 131865-50G
BT131865-250GEA 297.39 USD
BT131865-250G BT131865-50G
Lithium metaborate 98.0-102.0% ACS
Lithium metaborate

CAS: 13453-69-5; EC No: 236-631-5; MDL No: MFCD00011089 Powder; Linear Formula: LiBO2; MW: 49.75 Melting Point: 845° Hygroscopic


  • Hygroscopic
  • Powder

Formula: BLiO₂
Melting Pt: 845 °C
MDL Number: MFCD00011089
CAS Number: 13453-69-5
EINECS: 236-631-5

Order Now

Specification Test Results

Assay 98.0-102.0%
Insoluble matter ≤0.01%
Ammonium (NH4) ≤0.01%
Nitrate and Nitrite (NO4, NO3) ≤0.003%
Sulfate (SO4) ≤0.01%
Iron (Fe) ≤10ppm
Copper (Cu) ≤10ppm
Lead (Pb) ≤50ppm
Zinc (Zn) ≤500ppm
Sodium (Na) ≤500ppm
Potassium ≤200ppm
Calcium (Ca) ≤100ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR