2-Bromo-3-chloropyridine ≥97%
Supplier: BeanTown Chemical
131845-1G
131845-5G
BT131845-1GEA
69
USD
BT131845-1G
BT131845-5G
2-Bromo-3-chloropyridine ≥97%
2-Bromo-3-chloropyridine
CAS: 96424-68-9; MDL No: MFCD04039344
Powder; Molecular Formula: C5H3BrClN ; MW: 192.44
Melting Point: 58-63°
- Powder
Formula:
C₅H₃BrClN Melting Pt: 58…63 °C |
MDL Number:
MFCD04039344 CAS Number: 96424-68-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...