D-(-)-Valine ≥98%
Supplier: BeanTown Chemical
131815-100G
131815-25G
131815-5G
BT131815-100GEA
366
USD
BT131815-100G
BT131815-25G
BT131815-5G
D-(-)-Valine ≥98%
D-(-)-Valine
CAS: 640-68-6; EC No: 211-368-9; MDL No: MFCD00064219; RTECS: YV9360000
Solid; Molecular Formula: C5H11NO2; MW: 117.15
Melting Point: 295° (sublimes)
- RTECS: YV9360000
- Solid
Formula:
C₅H₁₁NO₂ Melting Pt: 295 °C (subl.) |
MDL Number:
MFCD00064219 CAS Number: 640-68-6 EINECS: 211-368-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...