D-Alanine ≥99%

Supplier: BeanTown Chemical

Synonyms: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine

131805-100G 131805-25G 131805-5G
BT131805-100GEA 396.49 USD
BT131805-100G BT131805-25G BT131805-5G
D-Alanine ≥99%
D-Alanine

CAS: 338-69-2; EC No: 206-418-1; MDL No: MFCD00008077 Solid; Molecular Formula: C3H7NO2; MW: 89.09 Melting Point: 291° (decomposes)


  • Solid

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 291 °C (dec.)
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR