Sodium molybdate(VI), anhydrous

Supplier: BeanTown Chemical

Synonyms: Sodium molybdenum oxide, Sodium molybdate, Molybdic acid sodium salt, Molybdic acid, sodium salt, di-Sodium molybdate

131760-100G 131760-500G
BT131760-100GEA 62.19 USD
BT131760-100G BT131760-500G
Sodium molybdate(VI), anhydrous
Sodium molybdate(VI)

CAS: 7631-95-0; EC No: 231-551-7; MDL No: MFCD00003486; RTECS: QA5085000 Crystalline/Powder; Linear Formula: Na2MoO4; MW: 205.92 Melting Point: 687° Density (g/mL): 3.78; Refractive Index: 1.714 Hygroscopic


  • RTECS: QA5085000
  • Refractive index: 1.714
  • Hygroscopic
  • Crystalline/Powder

Formula: MoNa₂O₄
MW: 205.92
Melting Pt: 687
Density: 3.78 g/mL
MDL Number: MFCD00003486
CAS Number: 7631-95-0
EINECS: 231-551-7
Merck Index: 13,08718

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR