2-Methyl-4,6-dimethoxypyrimidine ≥97%

Supplier: BeanTown Chemical

131715-1G 131715-5G
BT131715-1GEA 86 USD
BT131715-1G BT131715-5G
2-Methyl-4,6-dimethoxypyrimidine ≥97%
2-Methyl-4,6-dimethoxypyrimidine

CAS: 13566-48-8; EC No: 254-941-9; MDL No: MFCD06657567 Solid; Molecular Formula: C7H10N2O2; MW: 154.17


  • Solid

Formula: C₇H₁₀N₂O₂
MW: 154.169 g/mol
MDL Number: MFCD06657567
CAS Number: 13566-48-8
EINECS: 254-941-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR