Potassium chloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Potassiumchloride
131700-1KG
131700-5KG
BT131700-1KGEA
49.83
USD
BT131700-1KG
BT131700-5KG
Potassium chloride ≥99%
Potassium chloride
CAS: 7447-40-7; EC No: 231-211-8; MDL No: MFCD00011360; RTECS: TS8050000
Crystalline/Powder; Linear Formula: KCl; MW: 74.55
Melting Point: 773°; Boiling Point: 1500°
Density (g/mL): 1.984; Refractive Index: 1.49
Hygroscopic
- RTECS: TS8050000
- Refractive index: 1.49
- Hygroscopic
- Crystalline/Powder
Formula:
KCl MW: 74.55 g/mol Boiling Pt: 1420 Melting Pt: 773 Density: 1.984 g/mL |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...