5-Amino-2,4,6-triiodoisophthalic acid ≥95%
Supplier: BeanTown Chemical
131685-10G
131685-250G
131685-50G
BT131685-10GEA
26
USD
BT131685-10G
BT131685-250G
BT131685-50G
5-Amino-2,4,6-triiodoisophthalic acid ≥95%
5-Amino-2,4,6-triiodoisophthalic acid
CAS: 35453-19-1; EC No: 252-575-4; MDL No: MFCD00190167
Powder; Molecular Formula: C8H4I3NO4; MW: 558.84
Melting Point: 265-270°
- Powder
Formula:
C₈H₄I₃NO₄ Melting Pt: 265…270 °C |
MDL Number:
MFCD00190167 CAS Number: 35453-19-1 EINECS: 252-575-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...