6-Chloro-7-iodo-7-deazapurine ≥96%
Supplier: BeanTown Chemical
Synonyms:
4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine
131675-1G
131675-5G
BT131675-1GEA
118
USD
BT131675-1G
BT131675-5G
6-Chloro-7-iodo-7-deazapurine ≥96%
6-Chloro-7-iodo-7-deazapurine
CAS: 123148-78-7; MDL No: MFCD09263258
Powder; Molecular Formula: C6H3ClIN3; MW: 279.47
Melting Point: 179-183° (decomposes)
Light Sensitive
- Light Sensitive
- Powder
Formula:
C₆H₃ClIN₃ MW: 279.47 g/mol Melting Pt: 179…183 °C (dec.) |
MDL Number:
MFCD09263258 CAS Number: 123148-78-7 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...