4-Cyanophenylhydrazine hydrochloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Hydrazinylbenzonitrile hydrochloride
131665-100G
131665-25G
131665-5G
BT131665-100GEA
727
USD
BT131665-100G
BT131665-25G
BT131665-5G
4-Cyanophenylhydrazine hydrochloride ≥97%
4-Cyanophenylhydrazine hydrochloride
CAS: 2863-98-1; MDL No: MFCD00673994
Powder; Molecular Formula: C7H7N3·HCl; MW: 169.61
Melting Point: 241-244° (decomposes)
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
C₇H₈ClN₃ MW: 169.61 g/mol Melting Pt: 241…244 °C (dec.) |
MDL Number:
MFCD00673994 CAS Number: 2863-98-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...