Titanium(II) oxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Titanium oxide, Titanium monoxide

131645-100G 131645-25G 131645-500G
BT131645-100GEA 279.76 USD
BT131645-100G BT131645-25G BT131645-500G
Titanium(II) oxide ≥99.9% (trace metals basis)
Titanium(II) oxide

CAS: 12137-20-1; EC No: 235-236-5; MDL No: MFCD00036279 Lump; Linear Formula: TiO; MW: 63.90 Melting Point: 1700°; Boiling Point: 3000° Density (g/mL): 4.95


  • Lump

Formula: OTi
MW: 63.88 g/mol
Boiling Pt: >3000 °C
Melting Pt: 1700 °C
Density: 4.95 g/mL
MDL Number: MFCD00036279
CAS Number: 12137-20-1
EINECS: 235-236-5

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR