N,N-Diethyl-p-phenylenediammonium sulfate ≥97%

Supplier: BeanTown Chemical

Synonyms: N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate

131615-1KG 131615-250G 131615-50G
BT131615-1KGEA 254 USD
BT131615-1KG BT131615-250G BT131615-50G
N,N-Diethyl-p-phenylenediammonium sulfate ≥97%
N,N-Diethyl-p-phenylenediammonium sulphate

CAS: 6283-63-2; EC No: 228-500-6; MDL No: MFCD00012993; RTECS: SS9625000 Solid; Linear Formula: (CH3CH2)2NC6H4NH2·H2SO4; Molecular Formula: C10H16N2·H2O4S; MW: 262.33 Melting Point: 184-186° Air Sensitive, Light Sensitive


  • RTECS: SS9625000
  • Air Sensitive, Light Sensitive
  • Solid

Formula: C₁₀H₁₈N₂O₄S
MW: 262.33 g/mol
Melting Pt: 184…186 °C
MDL Number: MFCD00012993
CAS Number: 6283-63-2
EINECS: 228-500-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR