5'-Deoxy-5-fluorocytidine

Supplier: BeanTown Chemical

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoropyrimidin-2(1H)-one

131565-1G 131565-5G
BT131565-1GEA 125 USD
BT131565-1G BT131565-5G
5'-Deoxy-5-fluorocytidine
5'-Deoxy-5-fluorocytidine

CAS: 66335-38-4; MDL No: MFCD07369278 Solid; Molecular Formula: C9H12FN3O4; MW: 245.21 Melting Point: 196-198°


  • Solid

Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
Melting Pt: 196…198 °C
MDL Number: MFCD07369278
CAS Number: 66335-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR