2-(Cyclohexylamino)ethanesulfonic acid (CHES) ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-Cyclohexyltaurine, CHES
131550-100G
131550-25G
131550-500G
BT131550-100GEA
134.84
USD
BT131550-100G
BT131550-25G
BT131550-500G
2-(Cyclohexylamino)ethanesulfonic acid (CHES) ≥99%
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
CAS: 103-47-9; EC No: 203-115-6; MDL No: MFCD00003835
Crystalline; Molecular Formula: C8H17NO3S; MW: 207.29
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₈H₁₇NO₃S MW: 207.29 g/mol |
MDL Number:
MFCD00003835 CAS Number: 103-47-9 EINECS: 203-115-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...