Ethyl oleate ≥70%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
131540-1KG
131540-250G
131540-5KG
BT131540-1KGEA
97
USD
BT131540-1KG
BT131540-250G
BT131540-5KG
Ethyl oleate ≥70%, Technical Grade
Ethyl oleate
CAS: 111-62-6; EC No: 203-889-5; MDL No: MFCD00009579; RTECS: RG3715000
Liquid; Molecular Formula: C20H38O2; MW: 310.51
Melting Point: -32°; Boiling Point: 205-208°; Flash point:
- RTECS: RG3715000
- Refractive index: 1.451
- Light Sensitive
- Liquid
Formula:
C₂₀H₃₈O₂ MW: 310.52 g/mol Boiling Pt: 205…208 °C Melting Pt: –32 °C Density: 0.87 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00009579 CAS Number: 111-62-6 EINECS: 203-889-5 |
Specification Test Results
Assay | 70% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...