Boc-cycloleucine ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-(Boc-amino)cyclopentanecarboxylic acid, 1-N-BOC-Aminocyclopentanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

131505-1G 131505-5G
BT131505-1GEA 66 USD
BT131505-1G BT131505-5G
Boc-cycloleucine ≥98%
Boc-cycloleucine

CAS: 35264-09-6; MDL No: MFCD01076126 Powder; Molecular Formula: C11H19NO4 ; MW: 229.27


  • Powder

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD01076126
CAS Number: 35264-09-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR