Dichloro(1,5-cyclooctadiene)platinum(II)
Supplier: BeanTown Chemical
131500-1G
131500-250MG
131500-5G
BT131500-1GEA
218
USD
BT131500-1G
BT131500-250MG
BT131500-5G
Dichloro(1,5-cyclooctadiene)platinum(II)
Dichloro(1,5-cyclooctadiene)platinum(II)
CAS: 12080-32-9; EC No: 235-144-5; MDL No: MFCD00012413
Solid; Molecular Formula: C8H12Cl2Pt; MW: 374.18
Melting Point: 285° (decomposes)
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₈H₁₂Cl₂Pt Melting Pt: 285 °C (dec.) |
MDL Number:
MFCD00012413 CAS Number: 12080-32-9 EINECS: 235-144-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...