7-Hydroxycoumarin ≥98%

Supplier: BeanTown Chemical

Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone

131480-100G 131480-25G
BT131480-100GEA 177 USD
BT131480-100G BT131480-25G
7-Hydroxycoumarin ≥98%
7-Hydroxycoumarin

CAS: 93-35-6; EC No: 202-240-3; MDL No: MFCD00006878; RTECS: GN6820000 Crystalline; Molecular Formula: C9H6O3; MW: 162.14 Melting Point: 230° (decomposes)


  • RTECS: GN6820000
  • Crystalline

Formula: C₉H₆O₃
MW: 162.14 g/mol
Melting Pt: 230 °C (dec.)
MDL Number: MFCD00006878
CAS Number: 93-35-6
EINECS: 202-240-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR