O-Benzyl-DL-serine

Supplier: BeanTown Chemical

Synonyms: H-DL-OBzl-Ser-OH, H-DL-Ser(Bzl)-OH

131475-1G 131475-5G
BT131475-1GEA 32 USD
BT131475-1G BT131475-5G
O-Benzyl-DL-serine
O-Benzyl-DL-serine

CAS: 5445-44-3; EC No: 226-650-7; MDL No: MFCD00021724 Solid; Molecular Formula: C10H13NO3; MW: 195.22 Melting Point: 225° (decomposes)


  • Solid

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 225 °C (dec.)
MDL Number: MFCD00021724
CAS Number: 5445-44-3
EINECS: 226-650-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR