4-Bromo-2-methylpyridine ≥97%

Supplier: BeanTown Chemical

131450-1G 131450-250MG
BT131450-1GEA 138 USD
BT131450-1G BT131450-250MG
4-Bromo-2-methylpyridine ≥97%
4-Bromo-2-methylpyridine

CAS: 22282-99-1; MDL No: MFCD03788196 Liquid; Molecular Formula: C6H6BrN; MW: 172.02 Flash point: 78°C (172°F) Density (g/mL): 1.45; Refractive Index: 1.556


  • Refractive index: 1.556
  • Liquid

Formula: C₆H₆BrN
Density: 1.45 g/mL
Flash Pt: 78 °C (172 °F)
MDL Number: MFCD03788196
CAS Number: 22282-99-1

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR