Dipraseodymium tricarbonate octahydrate ≥99.99% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Praseodymium(III) carbonate octahydrate
131425-100G
131425-25G
131425-5G
BT131425-100GEA
425
USD
BT131425-100G
BT131425-25G
BT131425-5G
Dipraseodymium tricarbonate octahydrate ≥99.99% (trace rare earth metals basis)
Dipraseodymium tricarbonate octahydrate
CAS: 14948-62-0; EC No: 227-578-9; MDL No: MFCD00211307
Crystalline; Linear Formula: Pr2(CO3)3·8H2O; MW: 605.96
- Crystalline
Formula:
C₃O₉Pr₂·8H₂O MW: 605.97 (461.85 anhy) |
MDL Number:
MFCD00211307 CAS Number: 14948-62-0 EINECS: 227-578-9 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...