Xylenol orange tetrasodium salt ACS

Supplier: BeanTown Chemical

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

131405-1G 131405-25G 131405-5G
BT131405-1GEA 26.47 USD
BT131405-1G BT131405-25G BT131405-5G
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt

CAS: 3618-43-7; EC No: 222-805-8; MDL No: MFCD00044293 Powder; Molecular Formula: C31H28N2Na4O13S; MW: 760.59 Melting Point: 195° (decomposes)


  • Powder

Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: 210 (dec.)
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR