6-Chloro-3-pyridineacetic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-(6-Chloropyridin-3-yl)acetic acid

131335-10G 131335-50G
BT131335-10GEA 76 USD
BT131335-10G BT131335-50G
6-Chloro-3-pyridineacetic acid ≥97%
6-Chloro-3-pyridineacetic acid

CAS: 39891-13-9; MDL No: MFCD01863172 Solid; Molecular Formula: C7H6ClNO2; MW: 171.58 Melting Point: 172-176°


  • Solid

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 172…176 °C
MDL Number: MFCD01863172
CAS Number: 39891-13-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR