2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

131295-1G 131295-25G 131295-5G
BT131295-1GEA 71 USD
BT131295-1G BT131295-25G BT131295-5G
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%
2,2'-Bipyridine-4,4'-dicarboxylic acid

CAS: 6813-38-3; MDL No: MFCD00015430 Crystalline; Molecular Formula: C12H8N2O4; MW: 244.20


  • Crystalline

Formula: C₁₂H₈N₂O₄
MW: 244.20 g/mol
MDL Number: MFCD00015430
CAS Number: 6813-38-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR