Lithium sulphate monohydrate ≥99.0% ACS

Supplier: BeanTown Chemical

131265-100G 131265-2KG 131265-500G
BT131265-100GEA 104.33 USD
BT131265-100G BT131265-2KG BT131265-500G
Lithium sulphate monohydrate ≥99.0% ACS
Lithium sulphate monohydrate

CAS: 10102-25-7; EC No: 233-820-4; MDL No: MFCD00149766 Crystalline; Linear Formula: Li2SO4·H2O ; MW: 127.96


  • Crystalline

Formula: Li₂SO₄·H₂O
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR