Succinic anhydride ≥99%

Supplier: BeanTown Chemical

Synonyms: Dihydrofuran-2,5-dione

131255-1KG 131255-500G
BT131255-1KGEA 53.78 USD
BT131255-1KG BT131255-500G
Succinic anhydride ≥99%
Succinic anhydride

CAS: 108-30-5; EC No: 203-570-0; MDL No: MFCD00005525; RTECS: WN0875000 Crystalline; Molecular Formula: C4H4O3; MW: 100.07 Melting Point: 118-120°; Boiling Point: 261°; Flash point: 157°C (314°F) Density (g/mL): 1.572 Moisture Sensitive


  • RTECS: WN0875000
  • Moisture Sensitive
  • Crystalline

Formula: C₄H₄O₃
MW: 100.07 g/mol
Boiling Pt: 261
Melting Pt: 120
Density: 1.572 g/mL
Flash Pt: 157 °C (314 °F)
MDL Number: MFCD00005525
CAS Number: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8 III
Merck Index: 12,09039

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR