Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥97%
Supplier: BeanTown Chemical
131245-1G
131245-250MG
BT131245-1GEA
306
USD
BT131245-1G
BT131245-250MG
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥97%
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
CAS: 25940-35-6; MDL No: MFCD01975194
Solid; Molecular Formula: C7H5N3O2; MW: 163.14
Melting Point: 275-280°
- Solid
Formula:
C₇H₅N₃O₂ Melting Pt: 275…280 °C |
MDL Number:
MFCD01975194 CAS Number: 25940-35-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...