Bromopentafluorobenzene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Bromo-2,3,4,5,6-pentafluorobenzene

131225-100G 131225-25G 131225-500G
BT131225-100GEA 332 USD
BT131225-100G BT131225-25G BT131225-500G
Bromopentafluorobenzene ≥99%
Bromopentafluorobenzene

CAS: 344-04-7; EC No: 206-449-0; MDL No: MFCD00000287; RTECS: CY9040552 Liquid; Molecular Formula: C6BrF5; MW: 246.96 Melting Point: -31°; Boiling Point: 137°; Flash point: 87°C (189°F) Density (g/mL): 1.981; Refractive Index: 1.449


  • RTECS: CY9040552
  • Refractive index: 1.449
  • Liquid

Formula: C₆BrF₅
MW: 246.96 g/mol
Boiling Pt: 137 °C
Melting Pt: –31 °C
Density: 1.981 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00000287
CAS Number: 344-04-7
EINECS: 206-449-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR