2-Bromofluorene ≥95%
Supplier: BeanTown Chemical
131160-25G
131160-5G
BT131160-25GEA
153
USD
BT131160-25G
BT131160-5G
2-Bromofluorene ≥95%
2-Bromofluorene
CAS: 1133-80-8; EC No: 214-480-6; MDL No: MFCD00001115
Crystalline/Powder; Molecular Formula: C13H9Br; MW: 245.11
Melting Point: 112-114°; Boiling Point: 185°/135 mmHg
- Crystalline/Powder
Formula:
C₁₃H₉Br Boiling Pt: 185 °C/135 mmHg Melting Pt: 112…114 °C |
MDL Number:
MFCD00001115 CAS Number: 1133-80-8 EINECS: 214-480-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...