Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98%

Supplier: BeanTown Chemical

131150-100G 131150-25G 131150-5G
BT131150-100GEA 265 USD
BT131150-100G BT131150-25G BT131150-5G
Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98%
Glycinamide hydrochloride (H-Gly-NH2.HCl)

CAS: 1668-10-6; EC No: 216-789-1; MDL No: MFCD00013008 Crystalline; Linear Formula: NH2CH2CONH2·HCl ; Molecular Formula: C2H6N2O·HCl ; MW: 110.54 Melting Point: 204° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₂H₇ClN₂O
Melting Pt: 204 °C (dec.)
MDL Number: MFCD00013008
CAS Number: 1668-10-6
EINECS: 216-789-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR