Sand, quartz, washed
Supplier: BeanTown Chemical
131135-1KG
131135-5KG
BT131135-1KGEA
42.73
USD
BT131135-1KG
BT131135-5KG
Sand, quartz, washed
Sand, quartz
CAS: 14808-60-7; EC No: 238-878-4; MDL No: MFCD02100519; RTECS: VV7330000
Solid; Linear Formula: SiO2; MW: 60.08
Melting Point: 1710°
- RTECS: VV7330000
- Solid
Formula:
SiO₂ MW: 60.08 g/mol Melting Pt: 1710 °C |
MDL Number:
MFCD02100519 CAS Number: 14808-60-7 EINECS: 238-878-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...