Diammonium wolframate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

131110-10G 131110-250G 131110-50G
BT131110-10GEA 78.3 USD
BT131110-10G BT131110-250G BT131110-50G
Diammonium wolframate ≥99.99% (trace metals basis)
Diammonium wolframate

CAS: 11140-77-5; MDL No: MFCD00015956 Powder; Linear Formula: (NH4)2WO4 ; MW: 283.91


  • Powder

Formula: H₈N₂O₄W
MDL Number: MFCD00015956
CAS Number: 11140-77-5

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR