1-Bromo-2-chlorobenzene ≥98%

Supplier: BeanTown Chemical

131105-1KG 131105-250G 131105-50G
BT131105-1KGEA 644 USD
BT131105-1KG BT131105-250G BT131105-50G
1-Bromo-2-chlorobenzene ≥98%
1-Bromo-2-chlorobenzene

CAS: 694-80-4; EC No: 211-775-1; MDL No: MFCD00000532 Liquid; Molecular Formula: C6H4BrCl; MW: 191.45 Boiling Point: 204°; Flash point: 91°C (196°F) Density (g/mL): 1.638; Refractive Index: 1.582


  • Refractive index: 1.582
  • Liquid

Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 204 °C
Density: 1.638 g/mL
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR