Polyethylene oxide, M.W. > 5000000

Supplier: BeanTown Chemical

131100-100G 131100-500G
BT131100-100GEA 57.15 USD
BT131100-100G BT131100-500G
Polyethylene oxide, M.W. > 5000000
Polyethylene oxide

CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD00147992 Powder; Linear Formula: (-CH2CH2O-)n Melting Point: 175° Density (g/mL): 1.21

Formula: (-CH₂CH₂O-)n
Melting Pt: 175 °C
Density: 1.21 g/mL
MDL Number: MFCD00147970
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR