2,4,6-Triisopropylbenzenesulfonyl azide ≥98%
Supplier: BeanTown Chemical
131025-1G
131025-5G
BT131025-1GEA
58
USD
BT131025-1G
BT131025-5G
2,4,6-Triisopropylbenzenesulfonyl azide ≥98%
2,4,6-Triisopropylbenzenesulfonyl azide
CAS: 36982-84-0; MDL No: MFCD00859286
Solid; Molecular Formula: C15H23N3O2S; MW: 309.43
Melting Point: 39-44°
- Solid
Formula:
C₁₅H₂₃N₃O₂S MW: 309.428 g/mol Melting Pt: 39…44 °C |
MDL Number:
MFCD00859286 CAS Number: 36982-84-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...