Samarium(III) oxide ≥99.98%
Supplier: BeanTown Chemical
130985-100G
130985-25G
130985-500G
BT130985-100GEA
231.68
USD
BT130985-100G
BT130985-25G
BT130985-500G
Samarium(III) oxide ≥99.98%
Samarium(III) oxide
CAS: 12060-58-1; EC No: 235-043-6; MDL No: MFCD00011237
Powder; Linear Formula: Sm2O3; MW: 348.72
Melting Point: 2325°
Density (g/mL): 8.35; Refractive Index: 1.97
- Refractive index: 1.97
- Powder
Formula:
O₃Sm₂ MW: 348.72 g/mol Melting Pt: 2325 Density: 8.35 g/mL |
MDL Number:
MFCD00011237 CAS Number: 12060-58-1 EINECS: 235-043-6 Merck Index: 14,08347 |
Specification Test Results
Assay | 99.98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...