2,7-Dihydroxy-9-fluorenone

Supplier: BeanTown Chemical

Synonyms: 2,7-Dihydroxy-9H-fluoren-9-one

130980-1G 130980-5G
BT130980-1GEA 86 USD
BT130980-1G BT130980-5G
2,7-Dihydroxy-9-fluorenone
2,7-Dihydroxy-9-fluorenone

CAS: 42523-29-5; MDL No: MFCD00037141 Solid; Molecular Formula: C13H8O3; MW: 212.20


  • Solid

Formula: C₁₃H₈O₃
MDL Number: MFCD00037141
CAS Number: 42523-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR